missing translation for 'onlineSavingsMsg'
Learn More

Potassium pyrosulfate, pure, Thermo Scientific™

Product Code. 17545595 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
2.50kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17545595 500 g 500g
17535595 2.5 kg 2.50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17545595 Supplier Thermo Scientific Acros Supplier No. 472465000
  • ONLINE EXCLUSIVE PRICE
    62.65 €
    60.80 € / 500g
    Save 1.85 €
    2% Off
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7790-62-7
Molecular Formula K2O7S2
Molecular Weight (g/mol) 254.31
MDL Number 00011385
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
IUPAC Name dipotassium (sulfonatooxy)sulfonate
SMILES [K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O

Specifications

Melting Point 325°C
Color White
Physical Form Adhering Crystalline Powder
Quantity 500 g
Linear Formula K2S2O7
Merck Index 13-7749
Solubility Information May decompose in water
Packaging Plastic Bottle
Chemical Name or Material Potassium pyrosulfate
compliance-icons compliance-icons
Product Identifier
  • Potassium pyrosulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Potassium pyrosulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.