CAS RN 78-48-8
IUPAC Name:
1-{[bis(butylsulfanyl)phosphoryl]sulfanyl}butane
Synonyms:
de-green def defoliant caswell no. 864 epa pesticide chemical code 074801 butyphos DEF chemagro B-1776 fossfall butyl phosphorotrithioate ((bus)3PO) butifos
Molecular Weight (g/mol):
314.5
Molecular Formula:
C12H27OPS3
InChi Key:
ZOKXUAHZSKEQSS-UHFFFAOYSA-N
SMILES:
CCCCSP(=O)(SCCCC)SCCCC
1
–
5
of
5
results
1
1,2,4-Tributyl Phosphorotrithioate, TRC
CAS: 78-48-8 Molecular Formula: C12 H27 O P S3 Molecular Weight (g/mol): 314.51 Synonym: Butyl phosphorotrithioate (6CI),B 1,776,Butiphos,Butyl phosphorotrithioate ((BuS)3PO),Chemagro B 1776,DEF,DEF (defoliant),DEF 6,De-Green,Fos-Fall A,Fosfall,S,S,S-Tributyl phosphorotrithioate,S,S,S-Tributyl trithiophosphate,TBPT,TBTP,Tribufos IUPAC Name: 1-bis(butylsulfanyl)phosphorylsulfanylbutane SMILES: CCCCSP(=O)(SCCCC)SCCCC
5
Tribufos 100 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality