missing translation for 'onlineSavingsMsg'
Learn More

Tribufos in Methyl-tert.butylether 1000μg/mL, Fisher Chemical™

Product Code. 12916023 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12916023 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12916023 Supplier Fisher Chemical Supplier No. F113421

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 78-48-8
Molecular Formula C12H27OPS3
Concentration 1000 μg/mL
InChI Key ZOKXUAHZSKEQSS-UHFFFAOYSA-N
Type Organic Single-component Standard Solution
SMILES CCCCSP(=O)(SCCCC)SCCCC
IUPAC Name 1-bis(butylsulfanyl)phosphorylsulfanylbutane
Solvent or Matrix Methyl-tert.butylether
Molecular Weight (g/mol) 314.501
PubChem CID 5125
Quantity 1 mL
ChEBI CHEBI:38737
Chemical Name or Material Tribufos (DEF)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.