accessibility menu, dialog, popup

UserName

CH3H2CO

CAS RN 78-85-3

IUPAC Name: 2-methylprop-2-enal
Synonyms: methacrolein
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: STNJBCKSHOAVAJ-UHFFFAOYSA-N
SMILES: CC(=C)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
Percent Purity 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (7)
  • (1)