CAS RN 78186-34-2
CAS RN 78186-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bisantrene, MedChemExpress
MedChemExpress Bisantrene is a highly effective antitumor drug, targets eukaryotic type II topoisomerases.