accessibility menu, dialog, popup

UserName

CAS RN 79-44-7

H3CNCH3ClO

CAS RN 79-44-7

IUPAC Name: N,N-dimethylcarbamoyl chloride
Synonyms: dimethylcarbamyl chloride
Molecular Weight (g/mol): 107.54
Molecular Formula: C3H6ClNO
InChi Key: YIIMEMSDCNDGTB-UHFFFAOYSA-N
SMILES: CN(C)C(Cl)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)