missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcarbamoyl Chloride, TRC

Product Code. 30383178
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383178 5 g
30381318 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383178 Supplier Toronto Research Chemicals Supplier No. TRCD4712955G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-44-7
Molecular Formula C3 H6 Cl N O
Molecular Weight (g/mol) 107.54
Synonym (Dimethylamino)carbonyl Chloride, (N,N-Dimethylamino)carbonyl Chloride, 1-Chloro-N,N-dimethylformamide, Dimethylcarbamic Acid Chloride, Dimethylcarbamoyl Chloride, Dimethylcarbamyl Chloride, Dimethylchloroformamide, N,N-Dimethylcarbamic Acid Chloride, N,N-Dimethylcarbamic Chloride, N,N-Dimethylcarbamidoyl Chloride, N,N-Dimethylcarbamoyl Chloride, N,N-Dimethylcarbamyl Chloride, N,N-Dimethylchloroformamide, NSC 122671, Dimethylcarbamoyl Chloride
IUPAC Name N,N-dimethylcarbamoyl chloride
SMILES CN(C)C(=O)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H6ClNO/c1-5(2)3(4)6/h1-2H3
Formula Weight 107.0138
Chemical Name or Material Dimethylcarbamoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.