missing translation for 'onlineSavingsMsg'
Learn More

Dimethylcarbamyl chloride, 96%

Product Code. 11302146
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302146 25 g 25g
11312146 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302146 Supplier Thermo Scientific Alfa Aesar Supplier No. L03415.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 79-44-7 | C3H6ClNO | 107.537 g/mol

Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylcarbamyl chloride is used to make dyes and pharmaceuticals.

Solubility
Reacts with water.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-44-7
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00000635
InChI Key YIIMEMSDCNDGTB-UHFFFAOYSA-N
Synonym dimethylcarbamoyl chloride, dimethylcarbamyl chloride, dimethylcarbamic chloride, dimethylcarbamoylchloride, carbamic chloride, dimethyl, dimethylcarbamic acid chloride, dimethyl carbamoyl chloride, n,n-dimethylcarbamyl chloride, carbamoyl chloride, dimethyl, dimethylcarbamidoyl chloride
PubChem CID 6598
ChEBI CHEBI:82280
IUPAC Name N,N-dimethylcarbamoyl chloride
SMILES CN(C)C(=O)Cl

Specifications

Melting Point -33°C
Boiling Point 167°C to 168°C
Linear Formula (CH3)2NCOCl
Refractive Index 1.453
UN Number UN2262
Beilstein 878197
Solubility Information Reacts with water.
Density 1.171
Quantity 25 g
Flash Point 68°C (154°F)
Odor Unpleasant
Formula Weight 107.54
Sensitivity Moisture sensitive
Percent Purity 96%
Chemical Name or Material Dimethylcarbamyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.