CAS RN 794568-90-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
BAY 73-6691 racemate, MedChemExpress
MedChemExpress BAY 73-6691 racemate is a phosphodiesterase 9 inhibitor extracted from patent WO 2017070293 A1.