accessibility menu, dialog, popup

UserName

CAS RN 79479-07-5

(R)CH3OO(S)HNH3COHOOCH3CH3CH3

CAS RN 79479-07-5

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxybutanoate
Synonyms: methyl 2-[(tert-butoxycarbonyl)amino]-3-hydroxybutanoate
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: MZMWAPNVRMDIPS-UHFFFAOYNA-N
SMILES: COC(=O)C(NC(=O)OC(C)(C)C)C(C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)