missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-L-threonine methyl ester, 95%

Product Code. 15438108
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438108 25 g 25g
15428108 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438108 Supplier Thermo Scientific Alfa Aesar Supplier No. H61545.14
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79479-07-5 | C10H19NO5 | 233.26 g/mol

N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-L-threonine methyl ester is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°C. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79479-07-5
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.26
MDL Number MFCD00270479
InChI Key MZMWAPNVRMDIPS-RQJHMYQMSA-N
Synonym boc-thr-ome, 2s,3r-methyl 2-tert-butoxycarbonyl amino-3-hydroxybutanoate, boc-l-threonine methyl ester, n-boc-l-threonine methyl ester, n-tert-butoxycarbonyl-l-threonine methyl ester, n-tert-butoxycarbonyl threonine methyl ester, methyl 2s,3r-2-tert-butoxycarbonyl amino-3-hydroxybutanoate, methyl n-tert-butoxycarbonyl-l-threoninate, n-t-butoxycarbonyl-l-threonine methyl ester
PubChem CID 11368286
IUPAC Name methyl (2S,3R)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)[C@@H](C)O

Specifications

Boiling Point 206°C
Color Yellow
Refractive Index 1.45
Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 233.26
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material N-Boc-L-threonine methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.