accessibility menu, dialog, popup

UserName

CAS RN 80-58-0

CH3BrOHO

CAS RN 80-58-0

IUPAC Name: 2-bromobutanoic acid
Synonyms: 2-bromobutyric acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: YAQLSKVCTLCIIE-UHFFFAOYNA-N
SMILES: CCC(Br)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)