missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 552.3 2-Bromobutanoic Acid 10000 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30311609
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30311609

Brand: Dr. Ehrenstorfer DREGA09010313MB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-bromobutanoic acid
CAS 80-58-0
Analyte or Component Names 2-bromobutanoic acid
Shipping Condition Room Temperature
Molecular Formula C4 H7 Br O2
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Butanoic acid, 2-bromo-, Butyric acid, 2-bromo- (6CI,7CI,8CI), 2-Bromobutanoic acid, (RS)-2-Bromobutyric acid, (RS)-α-Bromobutyric acid, (±)-2-Bromobutanoic acid, (±)-2-Bromobutyric acid, (±)-α-Bromobutanoic acid, 2-Bromobutyric acid, DL-2-Bromo-n-butyric acid, DL-α-Bromobutyric acid, NSC 8024, α-Bromo-n-butyric acid, α-Bromobutyric acid
InChI Formula InChI=1S/C4H7BrO2/c1-2-3(5)4(6)7/h3H,2H2,1H3,(H,6,7)
SMILES CCC(Br)C(=O)O
IUPAC Name 2-bromobutanoic acid
Solvent or Matrix MTBE
Molecular Weight (g/mol) 167
Quantity 1 mL
Formula Weight 165.9629 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.