accessibility menu, dialog, popup

UserName

CH3ClSOOCl

CAS RN 80563-86-6

IUPAC Name: 3-chloro-2-methylbenzene-1-sulfonyl chloride
Synonyms: 3-chloro-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 225.08
Molecular Formula: C7H6Cl2O2S
InChi Key: ZSIYKAQPQRTBPF-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1Cl)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)