missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-2-methylbenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 10763455 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763455 5 g Glass bottle 5g
10487392 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10763455 Supplier Thermo Scientific Acros Supplier No. 357080050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 80563-86-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.09
MDL Number MFCD00052198
InChI Key ZSIYKAQPQRTBPF-UHFFFAOYSA-N
Synonym 3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
PubChem CID 522734
IUPAC Name 3-chloro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC=C1Cl)S(=O)(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 71°C to 73°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula Cl(CH3)C6H3SO2Cl
Quantity 5 g
Formula Weight 225.09
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 3-chloro-2-methylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-2-methylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.