missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzenesulfonyl chloride, 95%, Thermo Scientific™

Product Code. 10452192
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10452192 10 g 10g
10205674 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10452192 Supplier Thermo Scientific Maybridge Supplier No. TL00453EA
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 80563-86-6
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
InChI Key ZSIYKAQPQRTBPF-UHFFFAOYSA-N
Synonym 3-chloro-2-methylbenzene-1-sulfonyl chloride, 3-chloro-2-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-methyl-9ci, 3-chloro-2-methylphenylsulfonyl chloride, 3-chloro-2-methyl-benzenesulfonyl chloride, chloro 3-chloro-2-methylphenyl sulfone, pubchem5096, acmc-209pjq, buttpark 3711-61, 3-chloro-2-methylbenzenesulfonylchloride
PubChem CID 522734
IUPAC Name 3-chloro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=CC=C1Cl)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Linear Formula Cl(CH3)C6H3SO2Cl
Quantity 10 g
Formula Weight 225.09
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-methylbenzene-1-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.