accessibility menu, dialog, popup

UserName

CAS RN 82-76-8

IUPAC Name: 8-(phenylamino)naphthalene-1-sulfonic acid
Synonyms: phenyl-peri acid ANS anilinonaphthalenesulfonic acid phenylperi acid peri acid, phenyl-
Molecular Weight (g/mol): 299.34
Molecular Formula: C16H13NO3S
InChi Key: FWEOQOXTVHGIFQ-UHFFFAOYSA-N
SMILES: OS(=O)(=O)C1=C2C(NC3=CC=CC=C3)=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (6)
  • (7)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)