missing translation for 'onlineSavingsMsg'
Learn More

8-Anilino-1-naphthalene sulfonic acid, Free acid, MP Biomedicals™

Product Code. 11421972 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11421972 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11421972 Supplier MP Biomedicals Supplier No. 0210563305
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 82-76-8
Molecular Formula C16H13NO3S
Quantity 5 g
Synonym 8-anilino-1-naphthalenesulfonic acid, phenylperi acid, n-phenyl peri acid, 8-phenylamino naphthalene-1-sulfonic acid, 1-anilino-8-naphthalenesulfonate, 1-anilino-8-naphthalenesulfonic acid, peri acid, phenyl, 1-naphthalenesulfonic acid, 8-phenylamino, 1-anilinonaphthalene-8-sulfonic acid, 8-anilino-1-naphthalene sulfonic acid
InChI Key FWEOQOXTVHGIFQ-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)NC2=CC=CC3=C2C(=CC=C3)S(=O)(=O)O
IUPAC Name 8-anilinonaphthalene-1-sulfonic acid
Molecular Weight (g/mol) 299.344
PubChem CID 1369
ChEBI CHEBI:39708
Formula Weight 299.3
Chemical Name or Material 8-Anilino-1-naphthalene sulfonic acid, Free Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.