accessibility menu, dialog, popup

UserName

ONH2HCl

CAS RN 83708-39-8

IUPAC Name: 3-phenoxypropan-1-amine hydrochloride
Synonyms: 3-phenoxypropan-1-amine hydrochloride
Molecular Weight (g/mol): 187.67
Molecular Formula: C9H14ClNO
InChi Key: JTWGNQQRTPIYSJ-UHFFFAOYSA-N
SMILES: Cl.NCCCOC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)