CAS RN 84222-50-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Amino-1,7-dihydro-7-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6H-purin-6-one, TRC
CAS: 84222-50-4 Molecular Formula: C9 H13 N5 O4 Molecular Weight (g/mol): 255.23 Synonym: 2-Amino-7-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-1,7-dihydro-6H-purin-6-one,Ganciclovir Imp. H (EP),N-7 Isomer of Ganciclovir IUPAC Name: 2-amino-7-(1,3-dihydroxypropan-2-yloxymethyl)-1H-purin-6-one SMILES: NC1=Nc2ncn(COC(CO)CO)c2C(=O)N1
2
2-Amino-7-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-1,7-dihydro-6H-purin-6-one (N-7 Isomer of Ganciclovir), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.