CAS RN 84468-24-6
IUPAC Name:
5-(piperazine-1-sulfonyl)isoquinoline
Synonyms:
5-(piperazine-1-sulfonyl)isoquinoline
Molecular Weight (g/mol):
277.34
Molecular Formula:
C13H15N3O2S
InChi Key:
UPTYCYWTFGTCCG-UHFFFAOYSA-N
SMILES:
O=S(=O)(N1CCNCC1)C1=C2C=CN=CC2=CC=C1
1
–
1
of
1
results
1
HA-100, MedChemExpress
MedChemExpress HA-100 is a potent protein kinase inhibitor, with IC50s of 4 μM, 8 μM, 12 μM and 240 μM for cGMP-dependent protein kinase (PKG), cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and MLC-kinase, respectively. HA-100 also used as a ROCK inhibitor.