missing translation for 'onlineSavingsMsg'
Learn More

HA-100, MedChemExpress

Product Code. 16473228
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473228 10 mg 10mg
16472676 25 mg 25mg
16469776 50 mg 50mg
16465957 5 mg 5mg
16454496 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473228 Supplier Medchem Express Supplier No. HY100984

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HA-100 is a potent protein kinase inhibitor, with IC50s of 4 μM, 8 μM, 12 μM and 240 μM for cGMP-dependent protein kinase (PKG), cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and MLC-kinase, respectively. HA-100 also used as a ROCK inhibitor.

HA-100 is a potent protein kinase inhibitor, with IC50s of 4 μM, 8 μM, 12 μM and 240 μM for cGMP-dependent protein kinase (PKG), cAMP-dependent protein kinase (PKA), protein kinase C (PKC) and MLC-kinase, respectively. HA-100 also used as a ROCK inhibitor.

Specifications

Chemical Name or Material HA-100
CAS 84468-24-6
Color White
Purity Grade Notes Research
Molecular Formula C13H15N3O2S
Quantity 10 mg
Solubility Information DMSO : 150 mg/mL (540.85 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=CC2=C1C=CN=C2)(N3CCNCC3)=O
Molecular Weight (g/mol) 277.34
Formula Weight 277.34
Percent Purity 99.77%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.