CAS RN 84777-06-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Phthalic acid, n-pentyl-isopentyl ester (mixture of isomers), Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
1,2-Benzenedicarboxylic Acid, Dipentylester, Branched and Linear, TRC
CAS: 84777-06-0 Molecular Formula: C18 H26 O4 Molecular Weight (g/mol): 306.4 Synonym: 1,2-Benzenedicarboxylic acid, dipentyl ester, branched and linear,Dipentyl phthalate branched and linear,Phthalic acid, n-pentyl-isopentyl ester (mixture of isomers) IUPAC Name: dipentan-2-yl benzene-1,2-dicarboxylate SMILES: CCCC(C)OC(=O)c1ccccc1C(=O)OC(C)CCC