accessibility menu, dialog, popup

UserName

CAS RN 849061-97-8

OOOHOHHOHOHO

CAS RN 849061-97-8

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one hydrate
Synonyms: quercetin hydrate
Molecular Weight (g/mol): 320.25
Molecular Formula: C15H12O8
InChi Key: OKXFBEYCJRMINR-UHFFFAOYSA-N
SMILES: O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)