missing translation for 'onlineSavingsMsg'
Learn More

Quercetin Hydrate, TRC

Product Code. 30133416
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133416 5 g 5g
30004564 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133416 Supplier Toronto Research Chemicals Supplier No. TRCQ5097035G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849061-97-8
Molecular Formula C15H10O7 • x(H2O)
Molecular Weight (g/mol) 302.241802
Synonym 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one Hydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-benzopyran-4-one Hydrate, 3,3',4',5,7-Pentahydroxyflavone, 3,5,7,3',4'-Pentahydroxyflavone Hydrate, 3,5,7-Trihydroxy-2-(3,4-dihydroxyphenyl)-4H-chromen-4-one Hydrate, 3'-Hydroxykaempferol Hydrate, C.I. 75670 Hydrate, Corvitin Hydrate, Cyanidelonon 1522 Hydrate, Korvitin Hydrate, LDN 0052529 Hydrate, Lipoflavon Hydrate, Meletin Hydrate, NSC 57655 Hydrate, NSC 9219 Hydrate, Quercetin Hydrate, Quercetine Hydrate, Quercetol Hydrate, Quertin Hydrate, Quertine Hydrate, Sophoretin Hydrate, Xanthaurine Hydrate, 3,4',5,5',7-Pentahydroxy-flavone Hydrate
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
SMILES O=C1C(O)=C(C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C13

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H10O7.H2O/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6;/h1-5,16-19,21H;1H2
Formula Weight 320.0532
Chemical Name or Material Quercetin Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.