accessibility menu, dialog, popup

UserName

CAS RN 850568-03-5

IUPAC Name: [2-(aminomethyl)-5-fluorophenyl]boronic acid hydrochloride
Synonyms: 2-(aminomethyl)-5-fluorophenylboronic acid hydrochloride
Molecular Weight (g/mol): 205.42
Molecular Formula: C7H10BClFNO2
InChi Key: URBGTEWFXWJWPS-UHFFFAOYSA-N
SMILES: Cl.NCC1=C(C=C(F)C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results