CAS RN 85943-26-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-tert-Butyl-2-methoxybenzaldehyde, TRC
CAS: 85943-26-6 Molecular Formula: C12H16O2 Molecular Weight (g/mol): 192.25 Synonym: 5-(1,1-Dimethylethyl)-2-methoxybenzaldehyde; IUPAC Name: 5-tert-butyl-2-methoxybenzaldehyde SMILES: COc1ccc(cc1C=O)C(C)(C)C