CAS RN 861875-60-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
NVP-BAW2881, MedChemExpress
MedChemExpress NVP-BAW2881 (BAW2881) is a potent and selective VEGFR2 inhibitor with an IC50 of 4 nM.