missing translation for 'onlineSavingsMsg'
Learn More

NVP-BAW2881, MedChemExpress

Product Code. 16465727
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465727 5 mg 5mg
16449556 50 mg 50mg
16442988 10 mg 10mg
16420016 2 mg 2mg
16404306 100 mg 100mg
16402576 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16465727 Supplier Medchem Express Supplier No. HY100394
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress NVP-BAW2881 (BAW2881) is a potent and selective VEGFR2 inhibitor with an IC50 of 4 nM.

NVP-BAW2881 (BAW2881) is a potent and selective VEGFR2 inhibitor with an IC50 of 4 nM.

Specifications

Chemical Name or Material NVP-BAW2881
CAS 861875-60-7
Color White
Purity Grade Notes Research
Molecular Formula C22H15F3N4O2
Quantity 5 mg
Synonym BAW2881
Solubility Information DMSO : ≥ 33 mg/mL (77.76 mM)
SMILES O=C(C1=C2C=CC(OC3=NC(N)=NC=C3)=CC2=CC=C1)NC4=CC=CC(C(F)(F)F)=C4
Molecular Weight (g/mol) 424.38
Formula Weight 424.38
Percent Purity 98.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.