CAS RN 862189-96-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mirodenafil dihydrochloride, MedChemExpress
MedChemExpress Mirodenafil dihydrochloride (SK3530 dihydrochloride) is a phosphodiesterase type 5 (PDE-5) inhibitor developed for the treatment of erectile dysfunction.