missing translation for 'onlineSavingsMsg'
Learn More

Mirodenafil dihydrochloride, MedChemExpress

Product Code. 16488847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488847 5 mg 5mg
16486258 10 mg 10mg
16442387 50 mg 50mg
16406606 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488847 Supplier Medchem Express Supplier No. HY14930A
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mirodenafil dihydrochloride (SK3530 dihydrochloride) is a phosphodiesterase type 5 (PDE-5) inhibitor developed for the treatment of erectile dysfunction.

Mirodenafil dihydrochloride (SK3530 dihydrochloride) is a phosphodiesterase type 5 (PDE-5) inhibitor developed for the treatment of erectile dysfunction.

Specifications

Chemical Name or Material Mirodenafil dihydrochloride
CAS 862189-96-6
Color White
Purity Grade Notes Research
Molecular Formula C26H39Cl2N5O5S
Quantity 5 mg
Synonym SK-3530 dihydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (165.40 mM)
SMILES [H]Cl.[H]Cl.O=C1C(N(CC)C=C2CCC)=C2N=C(C3=CC(S(=O)(N4CCN(CCO)CC4)=O)=CC=C3OCCC)N1
Molecular Weight (g/mol) 604.59
Formula Weight 604.59
Percent Purity 99.79%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.