CAS RN 863513-93-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cebranopadol ((1α,4α)stereoisomer), MedChemExpress
MedChemExpress Cebranopadol ((1α,4α)stereoisomer) is a stereoisomer of cebranopadol. Cebranopadol is a potent agonist activity on ORL-1.