missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cebranopadol ((1α,4α)stereoisomer), MedChemExpress
Description
Cebranopadol ((1α,4α)stereoisomer) is a stereoisomer of cebranopadol. Cebranopadol is a potent agonist activity on ORL-1.
Specifications
Specifications
| Chemical Name or Material | Cebranopadol ((1α,4α)stereoisomer) |
| CAS | 863513-93-3 |
| Color | Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C24H27FN2O |
| Quantity | 2 mg |
| Synonym | GRT6005 (1α,4α)stereoisomer |
| Solubility Information | H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble) |
| SMILES | CN(C)[C@@]1(C2=CC=CC=C2)CC[C@]3(C(N4)=C(CCO3)C5=C4C=CC(F)=C5)CC1 |
| Molecular Weight (g/mol) | 378.48 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?