CAS RN 863774-58-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ Dimethylenastron
Inhibitor of mitotic motor kinesin Eg5
2
Dimethylenastron, MedChemExpress
MedChemExpress Dimethylenastron is a potent kinesin Eg5 inhibitor, with an IC50 of 200 nM.
3
4