CAS RN 863780-45-4
IUPAC Name:
magnesium(2+) 4-(2-methylpropyl)benzen-1-ide bromide
Synonyms:
magnesium(2+) 4-(2-methylpropyl)benzen-1-ide bromide
Molecular Weight (g/mol):
237.42
Molecular Formula:
C10H13BrMg
InChi Key:
VMAJYUMMJYAXIG-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CC(C)CC1=CC=[C-]C=C1