missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylphenylmagnesium bromide, 0.5M in 2-MeTHF

Product Code. 15460886
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460886 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15460886 Supplier Thermo Scientific Alfa Aesar Supplier No. H54201.AE
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 863780-45-4 | C10H13BrMg | 237.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 863780-45-4
Molecular Formula C10H13BrMg
MDL Number MFCD11113503
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym p-isobutylphenyl magnesium bromide, qoxyxvturbxklc-uhfffaoysa-m, grignard reagent
InChI Key VMAJYUMMJYAXIG-UHFFFAOYSA-M
SMILES CC(C)CC1=CC=[C-]C=C1.[Mg+2].[Br-]
IUPAC Name magnesium;2-methylpropylbenzene;bromide
Molecular Weight (g/mol) 237.423
PubChem CID 86067760
Formula Weight 237.42
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 4-Isobutylphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.