CAS RN 864082-47-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GSK429286A, MedChemExpress
MedChemExpress GSK429286A is a selective inhibitor of ROCK1 with an IC50 value of 14 nM.