missing translation for 'onlineSavingsMsg'
Learn More

GSK429286A, MedChemExpress

Product Code. 16485256
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485256 100 mg 100mg
16470667 50 mg 50mg
16457057 5 mg 5mg
16414348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485256 Supplier Medchem Express Supplier No. HY11000
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress GSK429286A is a selective inhibitor of ROCK1 with an IC50 value of 14 nM.

GSK429286A is a selective inhibitor of ROCK1 with an IC50 value of 14 nM.

Specifications

Chemical Name or Material GSK429286A
CAS 864082-47-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H16F4N4O2
Quantity 100 mg
Solubility Information DMSO : ≥ 51 mg/mL (117.95 mM)
SMILES O=C(C(C(C1=CC=C(C(F)(F)F)C=C1)C2)=C(C)NC2=O)NC3=CC4=C(NN=C4)C=C3F
Molecular Weight (g/mol) 432.37
Formula Weight 432.37
Percent Purity 98.15%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.