CAS RN 864864-86-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DFMTI, MedChemExpress
MedChemExpress DFMTI can completely block the rmGlu1 L757V glutamate response.