missing translation for 'onlineSavingsMsg'
Learn More

DFMTI, MedChemExpress

Product Code. 16490639
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490639 1 mg 1mg
16454306 100 mg 100mg
16445737 5 mg 5mg
16432998 10 mg 10mg
16429566 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490639 Supplier Medchem Express Supplier No. HY100404
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DFMTI can completely block the rmGlu1 L757V glutamate response.

DFMTI can completely block the rmGlu1 L757V glutamate response.

Specifications

Chemical Name or Material DFMTI
CAS 864864-86-8
Color White
Purity Grade Notes Research
Molecular Formula C20H18F2N4O
Quantity 1 mg
Synonym MK5435
Solubility Information DMSO : 25 mg/mL (67.86 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N(C(C)C)C1)C2=C1C=C(C3=C(C)N(C4=CC=C(F)C=C4F)N=N3)C=C2
Molecular Weight (g/mol) 368.38
Formula Weight 368.38
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.