CAS RN 865608-11-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ CMPD101
Potent and selective GRK2/3 inhibitor
2
CMPD101, TRC
CAS: 865608-11-3 Molecular Formula: C24H21F3N6O Molecular Weight (g/mol): 466.46 Synonym: 3-[[[4-Methyl-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]methyl]amino]-N-[[2-(trifluoromethyl)phenyl]methyl]benzamide,Takeda101 IUPAC Name: 3-[(4-methyl-5-pyridin-4-yl-1,2,4-triazol-3-yl)methylamino]-N-[[2-(trifluoromethyl)phenyl]methyl]benzamide SMILES: O=C(NCC1=CC=CC=C1C(F)(F)F)C2=CC=CC(NCC3=NN=C(C4=CC=NC=C4)N3C)=C2
3