CAS RN 866405-64-3
IUPAC Name:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine
Synonyms:
4-(6-{4-[2-(piperidin-1-yl)ethoxy]phenyl}pyrazolo[1,5-a]pyrimidin-3-yl)pyridine
Molecular Weight (g/mol):
399.5
Molecular Formula:
C24H25N5O
InChi Key:
XHBVYDAKJHETMP-UHFFFAOYSA-N
SMILES:
C(CN1CCCCC1)OC1=CC=C(C=C1)C1=CN2N=CC(=C2N=C1)C1=CC=NC=C1
1
–
2
of
2
results
1
STEMCELL Technologies Dorsomorphin
BMP and AMPK pathway inhibitor; Inhibits ALK2, ALK3, ALK6, and AMPK
2
Dorsomorphin, MedChemExpress
MedChemExpress Dorsomorphin (Compound C) is a selective and ATP-competitive AMPK inhibitor (Ki=109 nM in the absence of AMP). Dorsomorphin (BML-275) selectively inhibits BMP type I receptors ALK2, ALK3, and ALK6. Dorsomorphin induces autophagy.