accessibility menu, dialog, popup

UserName

CH3H3CNNH3CH3CCH3H3CCH3Cl

CAS RN 866926-59-2

IUPAC Name: 3-[2,6-bis(propan-2-yl)phenyl]-1-(2,4,6-trimethylphenyl)-4,5-dihydro-3H-1λ⁵-imidazol-1-ylium chloride
Synonyms: 3-(2,6-diisopropylphenyl)-1-(2,4,6-trimethylphenyl)-4,5-dihydro-1λ⁵-imidazol-1-ylium chloride
Molecular Weight (g/mol): 384.99
Molecular Formula: C24H33ClN2
InChi Key: RHFFYNDQCMNENO-UHFFFAOYSA-M
SMILES: [Cl-].CC(C)C1=CC=CC(C(C)C)=C1N1CC[N+](=C1)C1=C(C)C=C(C)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results