missing translation for 'onlineSavingsMsg'
Learn More

1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, 97%, Thermo Scientific™

Product Code. 11936661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936661 500 mg 500mg
11926661 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936661 Supplier Thermo Scientific Acros Supplier No. 435105000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 866926-59-2
Molecular Formula C24H33N2·Cl
Molecular Weight (g/mol) 385
InChI Key RHFFYNDQCMNENO-UHFFFAOYSA-M
Synonym 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-imidazolinium chloride, 1-2,6-di-i-propylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydroimidazolium chloride, 1-2,6-diisopropylphenyl-3-mesityl-4,5-dihydro-1h-imidazol-3-ium chloride, 1-2,6-diisopropylphenyl-3-2,4,6-trimethylphenyl-4,5-dihydro-1??-imidazol-1-ylium chloride, 3-2,6-bis 1-methylethyl phenyl-4,5-dihydro-1-2,4,6-trimethylphenyl-1h-imidazolium chloride
PubChem CID 11617932
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazol-3-ium;chloride
SMILES CC1=CC(=C(C(=C1)C)[N+]2=CN(CC2)C3=C(C=CC=C3C(C)C)C(C)C)C.[Cl-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 318°C to 319°C
Color Violet to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 385
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-(2,6-Diisopropylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.