CAS RN 87-24-1
IUPAC Name:
ethyl 2-methylbenzoate
Synonyms:
ethyl 2-methylbenzoate
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
SOUAXOGPALPTTC-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CC=CC=C1C
1
–
1
of
1
results
1
Ethyl o-toluate, 98+%
CAS: 87-24-1 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00009112 InChI Key: SOUAXOGPALPTTC-UHFFFAOYSA-N Synonym: ethyl o-toluate,ethyl o-methylbenzoate,ethyl orthotoluate,2-methylbenzoic acid ethyl ester,benzoic acid, 2-methyl-, ethyl ester,ethyl-o-toluate,2-methyl-benzoic acid ethyl ester,o-toluic acid ethyl ester,ethyl-2-methylbenzoate,ethylo-toluate PubChem CID: 66598 IUPAC Name: ethyl 2-methylbenzoate SMILES: CCOC(=O)C1=CC=CC=C1C