missing translation for 'onlineSavingsMsg'
Learn More

Ethyl o-toluate, 98+%

Product Code. 11390124
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390124 100 g 100g
11380124 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11390124 Supplier Thermo Scientific Alfa Aesar Supplier No. L00598.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 87-24-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-24-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009112
InChI Key SOUAXOGPALPTTC-UHFFFAOYSA-N
Synonym ethyl o-toluate, ethyl o-methylbenzoate, ethyl orthotoluate, 2-methylbenzoic acid ethyl ester, benzoic acid, 2-methyl-, ethyl ester, ethyl-o-toluate, 2-methyl-benzoic acid ethyl ester, o-toluic acid ethyl ester, ethyl-2-methylbenzoate, ethylo-toluate
PubChem CID 66598
IUPAC Name ethyl 2-methylbenzoate
SMILES CCOC(=O)C1=CC=CC=C1C

Specifications

Density 1.033
Boiling Point 227°C
Flash Point 91°C (195°F)
Refractive Index 1.507
Quantity 100 g
Beilstein 2042517
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Ethyl o-toluate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.