accessibility menu, dialog, popup

UserName

OO

CAS RN 87-41-2

IUPAC Name: 1,3-dihydro-2-benzofuran-1-one
Synonyms: phthalide
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: WNZQDUSMALZDQF-UHFFFAOYSA-N
SMILES: O=C1OCC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (10)
  • (5)
Quantity 
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (5)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (3)
  • (3)
  • (3)
  • (5)
  • (10)
  • (5)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)