missing translation for 'onlineSavingsMsg'
Learn More

Phthalide, 99%

Product Code. 10795431 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795431 100 g Plastic bottle 100g
10785431 5 g Glass Bottle 5g
10163230 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10795431 Supplier Thermo Scientific Acros Supplier No. 131091000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-41-2 | C8H6O2 | 134.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-41-2
MDL Number MFCD00005906
InChI Key WNZQDUSMALZDQF-UHFFFAOYSA-N
Synonym phthalide, isobenzofuran-1 3h-one, 1 3h-isobenzofuranone, 1-phthalanone, phthalolactone, 2-benzofuran-1 3h-one, isobenzofuranone, phthalanone, 3h-isobenzofuran-1-one, 2-hydroxymethylbenzoic acid, gamma-lactone
PubChem CID 6885
ChEBI CHEBI:38085
IUPAC Name 3H-2-benzofuran-1-one
SMILES C1C2=CC=CC=C2C(=O)O1

Specifications

Melting Point 70°C to 73°C
Color White to Beige
Boiling Point 290°C
Flash Point 152°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula C8H6O2
Quantity 100 g
Beilstein 17,31
Fieser 05,537; 08,402
Solubility Information Solubility in water: sparingly soluble
Molecular Weight (g/mol) 134.13
Formula Weight 134.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phthalide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Phthalide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.