accessibility menu, dialog, popup

UserName

CH3H3CH2N

CAS RN 87-62-7

IUPAC Name: 2,6-dimethylaniline
Synonyms: 2,6-dimethylaniline vic-m-xylidine 2,6-xylidine, 3H-labeled 3h-labeled 2,6-xylidine o-xylidine 3H-labeled 2,6-xylidine 2,6-xylidine, 3h-labeled
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: UFFBMTHBGFGIHF-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1N
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (7)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)