accessibility menu, dialog, popup

UserName

BFOOHHOOH

CAS RN 871329-84-9

IUPAC Name: 3-(dihydroxyboranyl)-5-fluorobenzoic acid
Synonyms: 3-(dihydroxyboranyl)-5-fluorobenzoic acid
Molecular Weight (g/mol): 183.93
Molecular Formula: C7H6BFO4
InChi Key: APHPXZJIUTVQNC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC(F)=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results